Human TCF-4/TCF7L2 ELISA Kit
Catalog # ABCAAB309185-96TST
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:TCF-4/TCF7L2
- Description:Human TCF-4/TCF7L2 ELISA kit
- Sample type:Cell lysate
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Detection range:187,5 - 12000 pg/ml
- Storage temperature:4 °C
- Sensitivity:49,873 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Human TCF-4/TCF7L2 ELISA kit is a single-wash 90-min Simplestep used to quantify Human TCF-4/TCF7L2 with a sensitivity of 49.873 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.