Order Entry
Italy
ContactUsLinkComponent
Cyclic GMP ELISA Kit
Cyclic GMP ELISA Kit
Catalog # ABCAAB133026-96
Supplier:  Abcam
CAS Number:  
undefined
Cyclic GMP ELISA Kit
Catalog # ABCAAB133026-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Target protein:
    cGMP
  • Description:
    Cyclic GMP ELISA kit
  • Sample type:
    Saliva, urine, plasma, cell culture supernatant, serum
  • Time to Results:
    3 h 0 min
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Cyclic GMP ELISA kit is a Competitive ELISA kit for the measurement of Cyclic GMP in Saliva, Urine, plasma, cell culture supernatant, Serum samples.