Order Entry
Italy
ContactUsLinkComponent
Human Urokinase type Plasminogen Activator ELISA Kit
Human Urokinase type Plasminogen Activator ELISA Kit
Catalog # ABCAAB108917-96
Supplier:  Abcam
CAS Number:  
undefined
Human Urokinase type Plasminogen Activator ELISA Kit
Catalog # ABCAAB108917-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Human Urokinase type Plasminogen Activator ELISA kit
Specifications
About this item
Human Urokinase type Plasminogen Activator ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Urokinase type Plasminogen Activator in Human in urine, plasma, serum, milk samples.