Order Entry
Italy
ContactUsLinkComponent
Pig IL-17 ELISA Kit
Pig IL-17 ELISA Kit
Catalog # ABCAAB193732-96
Supplier:  Abcam
CAS Number:  
undefined
Pig IL-17 ELISA Kit
Catalog # ABCAAB193732-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Pig IL-17 ELISA Kit
  • Tests per kit:
    96 tests
Specifications
About this item
Pig IL-17 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Pig IL-17 in Horse, Cow, Pig in Plasma, Cell culture supernatant, Serum samples.