Order Entry
Italy
ContactUsLinkComponent
Human VEGF R1 (FLT1) Antibody Pair - BSA and Azide free
Human VEGF R1 (FLT1) Antibody Pair - BSA and Azide free
Catalog # ABCAAB253794-10X96
Supplier:  Abcam
CAS Number:  
undefined
Human VEGF R1 (FLT1) Antibody Pair - BSA and Azide free
Catalog # ABCAAB253794-10X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Human VEGF R1 (FLT1) antibody pair - BSA and Azide free
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    10×96 tests

Specifications

About this item

Human VEGF R1 (FLT1) Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human VEGF R1 (FLT1).