Human MSH2 ELISA Kit
Catalog # ABCAAB233629-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:MSH2
- Description:Human MSH2 ELISA kit
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Detection range:1,56 - 100 ng/ml
- Storage temperature:4 °C
- Sensitivity:69 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Human MSH2 ELISA kit is a single-wash 90-min Simplestep used to quantify Human MSH2 with a sensitivity of 69 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.