Human Dectin-1 ELISA Kit
Catalog # ABCAAB309390-96TST
Supplier: Abcam
New Product
CAS Number:
Specifications
- Description:Human Dectin-1 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Human Dectin-1 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Dectin-1 in Human in cell culture supernatant, plasma, serum samples.