Mouse Myosin light chain 3 Antibody Pair - BSA and Azide Free
ABCAAB288643-10X96
: Abcam
:
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:Myosin light chain 3
- Description:Mouse Myosin light chain 3 antibody pair - BSA and azide free
- Detection range:39,06 - 5000 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Mouse Myosin light chain 3 antibody pair - BSA and Azide free.