Mouse Interferon gamma ELISPOT Kits
Catalog # ABCAAB64029-96
Nome del fornitore: Abcam
Numero CAS:
Specifiche
- Description:Mouse Interferon gamma ELISPOT kit
- Seq:1
- Species reactivity:Mouse
- Form:Pre-coated
- Antigen name:Interferon gamma
- Storage temperature:4 °C
- Processing time:Multiple steps
- Regulatory status:RUO
- Tests per kit:96 tests
Specifiche
Informazioni su questo articolo
Mouse Interferon gamma ELISPOT Kit is a ELISPOT Kit for the sensitive detection of Mouse Interferon gamma production in a single cell suspension.