Order Entry
Italy
ContactUsLinkComponent
Mouse P-Selectin ELISA Kit (CD62P)
Catalog # ABCAAB200014-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse P-Selectin ELISA Kit (CD62P)
Catalog # ABCAAB200014-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse P-Selectin ELISA Kit (CD62P)
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse P-Selectin ELISA Kit (CD62P) is a single-wash 90-min Simplestep used to quantify Mouse P-Selectin (CD62P) with a sensitivity of 4,16 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.