Order Entry
Italy
ContactUsLinkComponent
Human IL-1 beta ELISA Set (without plates)
Human IL-1 beta ELISA Set (without plates)
Catalog # ABCAAB47351-480
Supplier:  Abcam
CAS Number:  
undefined
Human IL-1 beta ELISA Set (without plates)
Catalog # ABCAAB47351-480
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    IL-1 beta
  • Description:
    Human IL-1 beta ELISA Set (without plates)
  • Sample type:
    Plasma, cell culture supernatant, serum
  • Detection method:
    Colorimetric
  • Time to Results:
    3 h 45 min
  • Detection range:
    15,6 - 500 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    5 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    5×96 tests

Specifications

About this item

Human IL-1 beta ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-1 beta ELISA Set (without plates) in Human in plasma, cell culture supernatant, Serum samples.