Order Entry
Italy
ContactUsLinkComponent
Human Semaphorin 7A Antibody Pair - BSA and Azide free
Human Semaphorin 7A Antibody Pair - BSA and Azide free
Catalog # ABCAAB253329-10X96
Supplier:  Abcam
CAS Number:  
undefined
Human Semaphorin 7A Antibody Pair - BSA and Azide free
Catalog # ABCAAB253329-10X96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Human Semaphorin 7A Antibody Pair - BSA and Azide free
  • Tests per kit:
    10×96 tests
Specifications
About this item
Human Semaphorin 7A Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Semaphorin 7A.