Rat Fractalkine ELISA Kit (CX3CL1)
Codice catalogo ABCAAB100761-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat Fractalkine ELISA Kit (CX3CL1)
- Tests per kit:96 tests
Specifications
About this item
Rat Fractalkine ELISA Kit (CX3CL1) is a Sandwich (quantitative) ELISA kit for the measurement of Rat Fractalkine (CX3CL1) in Rat in Cell culture extracts, Tissue Extracts samples.