Cyclic GMP Complete ELISA Kit
Codice catalogo ABCAAB133052-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Cyclic GMP Complete ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Cyclic GMP Complete ELISA Kit is a Competitive ELISA kit for the measurement of Cyclic GMP Complete in Saliva, Urine, Plasma, Tissue, Cell culture supernatant, Serum, Cell Lysate samples.