Order Entry
Italy
ContactUsLinkComponent
SARS-CoV-2 Nucleoprotein Antibody Pair - BSA and Azide Free
SARS-CoV-2 Nucleoprotein Antibody Pair - BSA and Azide Free
Catalog # ABCAAB309552-10X96
Supplier:  Abcam
CAS Number:  
undefined
SARS-CoV-2 Nucleoprotein Antibody Pair - BSA and Azide Free
Catalog # ABCAAB309552-10X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Format:
    Uncoated
  • Target protein:
    SARS-CoV-2 Nucleoprotein
  • Description:
    SARS-CoV-2 Nucleoprotein antibody pair - BSA and azide free
  • Detection range:
    23,44 - 3000 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Tests per kit:
    10×96 tests

Specifications

About this item

SARS-CoV-2 Nucleoprotein antibody pair - BSA and Azide free.