Monkey PD-L1 [28-8] ELISA Kit
Catalog # ABCAAB279408-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Monkey PD-L1 [28-8] ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Monkey PD-L1 [28-8] ELISA Kit is a single-wash 90-min Simplestep used to quantify Monkey PD-L1 [28-8] with a sensitivity of 15,85 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.