8-hydroxy-2'-deoxyguanosine (8-OHdG) ELISA Kit
Catalog # ABCAAB201734-96
Supplier: Abcam
CAS Number:
Specifications
- Description:8-hydroxy-2'-deoxyguanosine (8-OHdG) ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
The 8-Hydroxy-2'-deoxyguanosine (8-OHdG) ELISA kit is a competitive ELISA designed to quantify 8-Hydroxy-2'-deoxyguanosine with a sensitivity of 0,59 ng/ml. - Marker for oxidative DNA damage - Wide dynamic range – quantifies 0,94 to 60 ng/ml - Compatible with multiple sample types – works with serum, plasma, saliva, urine, cell lysates, and tissue extracts - Cited in over 100 publications.