Order Entry
Italy
ContactUsLinkComponent
Human Interferon gamma + IL4 ELISPOT Kits
Human Interferon gamma + IL4 ELISPOT Kits
Catalog # ABCAAB46649-480
Supplier:  Abcam
CAS Number:  
undefined
Human Interferon gamma + IL4 ELISPOT Kits
Catalog # ABCAAB46649-480
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Human Interferon gamma + IL4 ELISPOT kit
  • Seq:
    1
  • Species reactivity:
    Human
  • Form:
    Pre-coated
  • Antigen name:
    Interferon gamma + IL4
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO
  • Tests per kit:
    5×96 tests

Specifications

About this item

Human Interferon gamma + IL4 ELISPOT Kit is a ELISPOT Kit for the sensitive detection of Human Interferon gamma + IL4 production in a single cell suspension.