Mouse IL-12 p40 ELISA Kit
Catalog # ABCAAB171179-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse IL-12 p40 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Mouse IL-12 p40 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IL-12 p40 in Mouse in cell culture supernatant, citrate plasma, EDTA plasma, heparin plasma, serum samples.