Mouse NOV ELISA Kit (CCN3)
Catalog # ABCAAB205570-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse NOV ELISA kit (CCN3)
Specifications
About this item
Mouse NOV ELISA kit (CCN3) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse NOV (CCN3) in Mouse in plasma, cell culture extracts, tissue extracts, cell culture supernatant samples.