Endothelin 1 ELISA Kit
ABCAAB133030-96
: Abcam
:
- Description:Endothelin 1 ELISA kit
- Tests per kit:96 tests
Endothelin 1 ELISA Kit ab133030 is used to quantify Endothelin 1 with a sensitivity of 0,41 pg/ml. Sandwich ELISA Kit.
Endothelin 1 ELISA Kit ab133030 is used to quantify Endothelin 1 with a sensitivity of 0,41 pg/ml. Sandwich ELISA Kit.