Order Entry
Italy
ContactUsLinkComponent
Mouse CXCL5 ELISA Kit (GCP-2)
Mouse CXCL5 ELISA Kit (GCP-2)
Catalog # ABCAAB100719-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse CXCL5 ELISA Kit (GCP-2)
Catalog # ABCAAB100719-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse CXCL5 ELISA kit (GCP-2)
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse CXCL5 ELISA Kit (GCP-2) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse CXCL5 (GCP-2) in Mouse in Plasma, Cell culture supernatant, Serum samples.