Human TNF alpha ELISPOT Set
Catalog # ABCAAB46550-480
Supplier: Abcam
CAS Number:
Specifications
- Description:Human TNF alpha ELISPOT set
- Seq:1
- Species reactivity:Human;Monkey
- Form:Uncoated
- Storage temperature:4 °C
- Processing time:Multiple steps
- Antigen symbol:TNF alpha
- Regulatory status:RUO
- Tests per kit:5×96 tests
Specifications
About this item
Human TNF alpha ELISPOT Set is a ELISPOT Set for the sensitive detection of Human TNF alpha production in a single cell suspension.