Human IFN gamma High Sensitivity ELISA Kit
Catalog # ABCAAB46048-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IFN gamma High Sensitivity ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human IFN gamma High Sensitivity ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IFN gamma High Sensitivity in Human in Plasma, Cell culture supernatant, Serum samples.