Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Assays
ELISAs
Rat FLT1 Antibody Pair - BSA and Azide free (VEGF-R1)
Rat FLT1 Antibody Pair - BSA and Azide free (VEGF-R1)
Rat FLT1 Antibody Pair - BSA and Azide free (VEGF-R1)
Catalog # ABCAAB253306-10X96
Supplier:
Abcam
CAS Number:
Expand image
Rat FLT1 Antibody Pair - BSA and Azide free (VEGF-R1)
Catalog # ABCAAB253306-10X96
Supplier:
Abcam
CAS Number:
Specifications
Description:
Rat FLT1 antibody pair - BSA and Azide free (VEGF-R1)
Tests per kit:
10×96 tests
Specifications
About this item
Product Details
Product Details