Monkey IL-10 ELISA Kit
Catalog # ABCAAB119530-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Monkey IL-10 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Monkey IL-10 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Monkey IL-10 in Monkey in Cell culture supernatant, Serum samples.