Order Entry
Italy
ContactUsLinkComponent
Human Interferon Gamma ELISPOT Kit
Human Interferon Gamma ELISPOT Kit
Catalog # ABCAAB62899-96
Supplier:  Abcam
CAS Number:  
undefined
Human Interferon Gamma ELISPOT Kit
Catalog # ABCAAB62899-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Human Interferon Gamma ELISPOT kit
  • Seq:
    1
  • Species reactivity:
    Human
  • Sample type:
    Suspension cells
  • Form:
    Pre-coated
  • Storage temperature:
    +4 °C
  • Processing time:
    Multiple steps
  • Antigen symbol:
    Interferon gamma
  • Regulatory status:
    RUO

Specifications

About this item

Human Interferon Gamma ELISPOT Kit is a ELISPOT Kit for the sensitive detection of Human Interferon Gamma production in a single cell suspension.