Order Entry
Italy
ContactUsLinkComponent
Human NOV ELISA Kit (Nephroblastoma Overexpressed)
Human NOV ELISA Kit (Nephroblastoma Overexpressed)
Catalog # ABCAAB193710-96
Supplier:  Abcam
CAS Number:  
undefined
Human NOV ELISA Kit (Nephroblastoma Overexpressed)
Catalog # ABCAAB193710-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Human NOV ELISA Kit (Nephroblastoma Overexpressed)
  • Tests per kit:
    96 tests
Specifications
About this item
Human NOV ELISA Kit (Nephroblastoma Overexpressed) is a Sandwich (quantitative) ELISA kit for the measurement of Human NOV (Nephroblastoma Overexpressed) in Human in Plasma, Cell culture supernatant, Serum samples.