Progesterone ELISA Kit
Catalog # ABCAAB285257-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Progesterone ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Progesterone ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Progesterone in Saliva, Serum, Tissue Culture Media, Plasma samples.