Order Entry
Italy
ContactUsLinkComponent
Pig Fibrinogen ELISA Kit
Pig Fibrinogen ELISA Kit
Catalog # ABCAAB205090-96
Supplier:  Abcam
CAS Number:  
undefined
Pig Fibrinogen ELISA Kit
Catalog # ABCAAB205090-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    Fibrinogen
  • Description:
    Pig Fibrinogen ELISA kit
  • Sample type:
    Citrate plasma, edta plasma, heparin plasma, serum
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 10 min
  • Detection range:
    12,5 - 400 ng/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    1,713 ng/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Pig Fibrinogen ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Pig Fibrinogen in Pig in Citrate plasma, EDTA plasma, heparin plasma, serum samples.