Mouse Relaxin 1 ELISA Kit
Catalog # ABCAAB213885-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Relaxin 1 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Mouse Relaxin 1 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Relaxin 1 in Mouse in Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum samples.