Order Entry
Italy
ContactUsLinkComponent
Human Tissue Plasminogen Activator ELISA Kit, Fluorescent
Human Tissue Plasminogen Activator ELISA Kit, Fluorescent
Catalog # ABCAAB282311-1X96
Supplier:  Abcam
CAS Number:  
undefined
Human Tissue Plasminogen Activator ELISA Kit, Fluorescent
Catalog # ABCAAB282311-1X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    Tissue Plasminogen Activator
  • Description:
    Human Tissue Plasminogen Activator ELISA kit, Fluorescent
  • Detection method:
    Fluorescent
  • Time to Results:
    1 h 30 min
  • Detection range:
    31,25 - 16000 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    13 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Human Tissue Plasminogen Activator ELISA kit, Fluorescent is a single-wash 90-min Simplestep used to quantify Human Tissue Plasminogen Activator with a sensitivity of 13 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Fluorescent Sandwich ELISA - 530/570/590 nm readout: works on any standard plate reader .