Order Entry
Italy
Orders LinkContactUsLinkComponent
Human GITR ELISA Kit

Human GITR ELISA Kit

Human GITR ELISA kit, 96 tests
Catalog # ABCAAB234571-96
Supplier:   Abcam
Human GITR ELISA Kit
Catalog # ABCAAB234571-96
Supplier:  Abcam
CAS Number:  
Human GITR ELISA Kit
Human GITR ELISA Kit

Human GITR ELISA kit, 96 tests

CAS Number:  
Price
769,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human GITR ELISA Kit is a single-wash 90-min Simplestep used to quantify Human GITR with a sensitivity of 14 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.

Specifications

Specifications

  • Description:
    Human GITR ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Cell culture extracts, Heparin Plasma, Citrate plasma, Serum, EDTA Plasma
  • Detection range:
    78 - 5000 pg/ml
  • Storage temperature:
    +4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB234571-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    14 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    GITR
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.