Canine IGFBP2 ELISA Kit
Catalog # ABCAAB273184-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Canine IGFBP2 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Canine IGFBP2 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Canine IGFBP2 in Dog in Plasma, Cell culture supernatant, Serum samples.