Order Entry
Italy
ContactUsLinkComponent
Overall Exosome Capture and Quantification ELISA Assay Kit
Overall Exosome Capture and Quantification ELISA Assay Kit
Catalog # ABCAAB285284-96
Supplier:  Abcam
CAS Number:  
undefined
Overall Exosome Capture and Quantification ELISA Assay Kit
Catalog # ABCAAB285284-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Overall Exosome Capture and Quantification ELISA Assay kit
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    96 tests

Specifications

About this item

Overall Exosome Capture and Quantification ELISA Assay kit is a Sandwich ELISA kit for the measurement of Overall Exosome Capture and Quantification ELISA Assay kit in plasma samples.