Order Entry
Italy
ContactUsLinkComponent
Mouse IL-2 ELISPOT Kit
Mouse IL-2 ELISPOT Kit
Catalog # ABCAAB64031-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse IL-2 ELISPOT Kit
Catalog # ABCAAB64031-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Mouse IL-2 ELISPOT kit
  • Seq:
    1
  • Species reactivity:
    Mouse
  • Sample type:
    Suspension cells
  • Form:
    Pre-coated
  • Storage temperature:
    +4 ℃
  • Processing time:
    Multiple steps
  • Antigen symbol:
    IL-2
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Mouse IL-2 ELISPOT kit is a ELISPOT kit for the sensitive detection of Mouse IL-2 production in a single cell suspension.