Mouse MMP-12 ELISA Kit
Catalog # ABCAAB213878-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse MMP-12 ELISA kit
Specifications
About this item
Mouse MMP-12 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse MMP-12 in Mouse in cell culture supernatant, Heparin plasma, serum samples.