Order Entry
Italy
Orders LinkContactUsLinkComponent
Human CD8A ELISA Kit

Human CD8A ELISA Kit

Human CD8A ELISA kit, 96 tests
Catalog # ABCAAB256393-1X96
Supplier:   Abcam
Human CD8A ELISA Kit
Catalog # ABCAAB256393-1X96
Supplier:  Abcam
CAS Number:  
Human CD8A ELISA Kit
Human CD8A ELISA Kit

Human CD8A ELISA kit, 96 tests

CAS Number:  
Price
759,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Human CD8A ELISA Kit is a single-wash 90-min Simplestep used to quantify Human CD8A with a sensitivity of 4,7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.

Specifications

Specifications

  • Description:
    Human CD8A ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Tissue Extracts, Cell culture supernatant, Serum, EDTA Plasma
  • Detection range:
    16 - 1000 pg/ml
  • Storage temperature:
    +4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB256393-1X96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    4,7 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    CD8 alpha
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.