Human Thrombin Antithrombin Complex total antigen assay ELISA Kit
Catalog # ABCAAB279724-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Pre-coated
- Target protein:Thrombin/Antithrombin complex
- Description:Human Thrombin Antithrombin Complex total antigen assay ELISA kit
- Detection method:Colorimetric
- Detection range:1 - 1000 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,718 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Human Thrombin Antithrombin Complex total antigen assay ELISA kit is a Quantitative ELISA kit for the measurement of Human Thrombin Antithrombin Complex total antigen assay in Human in Citrate plasma, EDTA plasma, Heparin plasma, plasma, Serum samples.