Human Kallikrein 13 ELISA Kit
Catalog # ABCAAB213803-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Kallikrein 13 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human Kallikrein 13 ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Human Kallikrein 13 in Human in cell lysate, cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum, Tissue Homogenate samples.