Mouse VEGF R3 ELISA Kit (FLT4)
Catalog # ABCAAB203367-96
Nome del fornitore: Abcam
Numero CAS:
Specifiche
- Description:Mouse VEGF R3 ELISA kit (FLT4)
- Tests per kit:96 tests
Specifiche
Informazioni su questo articolo
Mouse VEGF R3 ELISA Kit (FLT4) is a single-wash 90-min Simplestep used to quantify Mouse VEGF R3 (FLT4) with a sensitivity of 17,97 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.