Canine SCF ELISA Kit
ABCAAB273181-1X96
: Abcam
:
- Description:Canine SCF ELISA kit
- Tests per kit:96 tests
Canine SCF ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Canine SCF in Dog in Plasma, cell culture supernatant, serum samples.