Order Entry
Italy
ContactUsLinkComponent
Human Anti-Epstein Barr virus IgM ELISA Kit (EBV-VCA)
Human Anti-Epstein Barr virus IgM ELISA Kit (EBV-VCA)
Catalog # ABCAAB108732-96
Supplier:  Abcam
CAS Number:  
undefined
Human Anti-Epstein Barr virus IgM ELISA Kit (EBV-VCA)
Catalog # ABCAAB108732-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Format:
    Pre-coated
  • Target protein:
    Epstein-Barr Virus (VCA) IgM
  • Description:
    Human Anti-Epstein Barr virus IgM ELISA kit (EBV-VCA)
  • Sample type:
    Citrate plasma, heparin plasma, serum
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Human Anti-Epstein Barr virus IgM ELISA kit (EBV-VCA) is a Indirect ELISA kit for the measurement of Human Anti-Epstein Barr virus IgM (EBV-VCA) in Human in Citrate plasma, heparin plasma, serum samples.