Order Entry
Italy
ContactUsLinkComponent
Rat Kininogen ELISA Kit (KNG) ELISA Kit
Rat Kininogen ELISA Kit (KNG) ELISA Kit
Catalog # ABCAAB157742-96
Supplier:  Abcam
CAS Number:  
undefined
Rat Kininogen ELISA Kit (KNG) ELISA Kit
Catalog # ABCAAB157742-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    Kininogen
  • Description:
    Rat Kininogen ELISA kit (KNG) ELISA kit
  • Sample type:
    Plasma, serum
  • Detection method:
    Colorimetric
  • Detection range:
    6,25 - 400 ng/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    4,9707 ng/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Rat Kininogen ELISA kit (KNG) ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat Kininogen (KNG) ELISA kit in Rat in plasma, Serum samples.