cGMP ELISA Kit
Catalog # ABCAAB285321-96
Supplier: Abcam
CAS Number:
Specifications
- Description:cGMP ELISA kit
- Tests per kit:96 tests
Specifications
About this item
cGMP ELISA Kit is a Competitive ELISA kit for the measurement of cGMP in Plasma, Serum, Other biological fluids samples.