Phosphotyrosine ROS1 ELISA Kit
Catalog # ABCAAB279910-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Phosphotyrosine ROS1 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Phosphotyrosine ROS1 ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phosphotyrosine ROS1 in Human in Cell Lysate samples.