Rabbit Angiopoietin-1 ELISA Kit
Catalog # ABCAAB273247-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rabbit Angiopoietin-1 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Rabbit Angiopoietin-1 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rabbit Angiopoietin-1 in Rabbit in Plasma, Cell culture supernatant, Serum samples.