Order Entry
Italy
ContactUsLinkComponent
Canine IL8 ELISA Kit (Interleukin-8)
Canine IL8 ELISA Kit (Interleukin-8)
Catalog # ABCAAB155465-96
Supplier:  Abcam
CAS Number:  
undefined
Canine IL8 ELISA Kit (Interleukin-8)
Catalog # ABCAAB155465-96
Supplier:  Abcam
CAS Number:  
About this item
Canine IL8 ELISA kit (Interleukin-8) is a Sandwich (quantitative) ELISA kit for the measurement of Canine IL8 (Interleukin-8) in Dog in plasma, cell culture supernatant, serum samples.