Order Entry
Italy
ContactUsLinkComponent
Mouse/Rat Bcl-xL ELISA Kit
Mouse/Rat Bcl-xL ELISA Kit
Catalog # ABCAAB227899-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse/Rat Bcl-xL ELISA Kit
Catalog # ABCAAB227899-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse/Rat Bcl-xL ELISA Kit
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse/Rat Bcl-xL ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse/Rat Bcl-xL with a sensitivity of 28 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.