Order Entry
Italy
ContactUsLinkComponent
Human IL-1 alpha ELISA Set (without plates)
Human IL-1 alpha ELISA Set (without plates)
Catalog # ABCAAB47350-480
Supplier:  Abcam
CAS Number:  
undefined
Human IL-1 alpha ELISA Set (without plates)
Catalog # ABCAAB47350-480
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Human IL-1 alpha ELISA Set (without plates)
  • Seq:
    1
  • Tests per kit:
    5×96 tests
Specifications
About this item
Human IL-1 alpha ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-1 alpha ELISA Set (without plates) in Human in Plasma, Cell culture supernatant, Serum samples.